missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human CCL2/MCP-1 Quantikine ELISA Kit
Human CCL2/MCP-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
7735.00 SEK - 44075.00 SEK
Specifications
Assay Range | 31.20 - 2000pg/mL |
---|---|
Assay Sensitivity | 10pg/mL |
Conjugate | HRP |
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Product Type | Solid Phase Sandwich ELISA |
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Sufficient For | Price | Quantity & Availability | ||||
18711934
|
R&D Systems™
DCP00 |
1 Plate | Sufficient materials to run ELISAs on at least one 96-well plate |
7735.00 SEK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18782034
|
R&D Systems™
SCP00 |
6 Plates | Sufficient materials to run ELISAs on at least six 96-well plates |
44075.00 SEK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Human CCL2/MCP-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human CCL2/JE/MCP-1 in cell culture supernates (200 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul), urine (100 ul).Specifications
31.20 - 2000pg/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human MCP-1 | |
ELISA | |
Cell Culture Supernates (200uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL),Urine (100uL) |
10pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum, Urine | |
Human | |
6347 | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human CCL2/MCP-1 Quantikine ELISA Kit