missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FZD6 Partial ORF (NP_003497, 71 a.a. - 181 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4630.00 SEK - 7020.00 SEK

Specifications

Accession Number NP_003497
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 8323
Molecular Weight (g/mol) 37.95kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16118425
View Documents
Abnova™
H00008323-Q01.25ug
25 ÎĽg
7020.00 SEK
25µg
Please to purchase this item. Need a web account? Register with us today!
16108425
View Documents
Abnova™
H00008323-Q01.10ug
10 ÎĽg
4630.00 SEK
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene represents a member of the 'frizzled' gene family, which encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The protein encoded by this family member contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, and seven transmembrane domains, but unlike other family members, this protein does not contain a C-terminal PDZ domain-binding motif. This protein functions as a negative regulator of the canonical Wnt/beta-catenin signaling cascade, thereby inhibiting the processes that trigger oncogenic transformation, cell proliferation, and inhibition of apoptosis. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: PNIETFLCKAFVPTCIEQIHVVPPCRKLCEKVYSDCKKLIDTFGIRWPEELECDRLQYCDETVPVTFDPHTEFLGPQKKTEQVQRDIGFWCPRHLKTSGGQGYKFLGIDQC
Specifications

Specifications

NP_003497
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.95kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Hfz6
FZD6
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
8323
FZD6 (Human) Recombinant Protein (Q01)
PNIETFLCKAFVPTCIEQIHVVPPCRKLCEKVYSDCKKLIDTFGIRWPEELECDRLQYCDETVPVTFDPHTEFLGPQKKTEQVQRDIGFWCPRHLKTSGGQGYKFLGIDQC
RUO
FZD6
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Abnova™ Human FZD6 Partial ORF (NP_003497, 71 a.a. - 181 a.a.) Recombinant Protein with GST-tag at N-terminal

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.