missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant G-CSF, ACF
Granulocyte colony-stimulating factor, animal component-free

2986.20 SEK - 56666.25 SEK
Specifications
Name | Colony-stimulating factor 3, CSF-3, MGI-1G, Pluripoietin |
---|---|
Cell Type | Cancer Cells and Cell Lines; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neurons |
Purity or Quality Grade | ≥95% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17109661
![]() |
STEMCELL Technologies
78138.1 |
100 ÎĽg |
11693.70 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17119661
![]() |
STEMCELL Technologies
78138.2 |
1000 ÎĽg |
56666.25 SEK
1000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199651
![]() |
STEMCELL Technologies
78138 |
10 ÎĽg |
2986.20 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Granulocyte colony-stimulating factor (G-CSF) is a member of the CSF family of glycoproteins that regulate hematopoietic cell proliferation, differentiation, and function
- It is a key cytokine involved in the production of neutrophils and the stimulation of granulocyte colony formation from hematopoietic progenitor cells
- G-CSF causes a range of effects including a transient reduction of SDF-1 expression, the activation of metalloproteases that cleave VCAM-1, and the release of norepinephrine from the sympathetic nervous system, leading to the release or mobilization of hematopoietic stem cells from the bone marrow into the periphery
- The G-CSF receptor is expressed on a variety of hematopoietic cells, including myeloid-committed progenitor cells, neutrophils, granulocytes, and monocytes
Specifications
Colony-stimulating factor 3, CSF-3, MGI-1G, Pluripoietin | |
Cancer Cells and Cell Lines; Hematopoietic Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neurons | |
≥95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Human Recombinant G-CSF, ACF