Learn More
STEMCELL Technologies Human Recombinant IFN-gamma
Interferon-gamma
Brand:  STEMCELL Technologies 78020.2
Encompass_Preferred
 View more versions of this product
Product Code. 17199361
  • 14547.60 SEK / 1000µg
This item is not returnable. View return policy
Description
Description
  • Interferon-gamma (IFN-γ), also known as type II interferon, is produced by T and NK cells, and in smaller amounts by dendritic cells and macrophages
  • IFN-γ is controlled by cytokines such as IL-12 and IL-18 secreted in response to infection
  • IFN-γ binds to a receptor complex and initiates signal transduction via the JAK/STAT pathway; this culminates in the transcription and activation of many genes that control a diverse array of immunological functions
  • IFN-γ stimulates the antimicrobial and anti-tumor activity of macrophages, NK cells, and neutrophils by promoting the activation of microbial effector functions such as production of reactive oxygen species, nitric oxide, and complement
  • IFN-γ enhances MHC class I and II expression in dendritic cells and mononuclear phagocytes, as well as the production of IL-12 by dendritic cells
Specifications
Specifications
Interferon gamma, Type II interferon
1000 μg
B Cells; Hematopoietic Stem and Progenitor Cells; Intestinal Cells; Mesenchymal Stem and Progenitor Cells; T Cells
≥95%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Human Recombinant IFN-gamma