missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant IFN-gamma
Interferon-gamma

1366.20 SEK - 14547.60 SEK
Specifications
Name | Interferon gamma, Type II interferon |
---|---|
Cell Type | B Cells; Hematopoietic Stem and Progenitor Cells; Intestinal Cells; Mesenchymal Stem and Progenitor Cells; T Cells |
Purity or Quality Grade | ≥95% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16553871
![]() |
STEMCELL Technologies
78020.1 |
50 ÎĽg |
1366.20 SEK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17189361
![]() |
STEMCELL Technologies
78020 |
100 ÎĽg |
2596.05 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199361
![]() |
STEMCELL Technologies
78020.2 |
1000 ÎĽg |
14547.60 SEK
1000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Interferon-gamma (IFN-Îł), also known as type II interferon, is produced by T and NK cells, and in smaller amounts by dendritic cells and macrophages
- IFN-Îł is controlled by cytokines such as IL-12 and IL-18 secreted in response to infection
- IFN-Îł binds to a receptor complex and initiates signal transduction via the JAK/STAT pathway; this culminates in the transcription and activation of many genes that control a diverse array of immunological functions
- IFN-Îł stimulates the antimicrobial and anti-tumor activity of macrophages, NK cells, and neutrophils by promoting the activation of microbial effector functions such as production of reactive oxygen species, nitric oxide, and complement
- IFN-Îł enhances MHC class I and II expression in dendritic cells and mononuclear phagocytes, as well as the production of IL-12 by dendritic cells
Specifications
Interferon gamma, Type II interferon | |
B Cells; Hematopoietic Stem and Progenitor Cells; Intestinal Cells; Mesenchymal Stem and Progenitor Cells; T Cells | |
≥95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Human Recombinant IFN-gamma