missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant IL-10
Interleukin 10

2596.05 SEK - 68359.95 SEK
Specifications
Cell Type | Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Myeloid Cells; Neurons; T Cells |
---|---|
Purity or Quality Grade | ≥95% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17109381
![]() |
STEMCELL Technologies
78024.2 |
1000 ÎĽg |
68359.95 SEK
1000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17189371
![]() |
STEMCELL Technologies
78024 |
50 ÎĽg |
8243.10 SEK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199371
![]() |
STEMCELL Technologies
78024.1 |
10 ÎĽg |
2596.05 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Interleukin 10 (IL-10) is the founding member of the IL-10 family of class II cytokines
- All of the IL-10 cytokine family members have a four-helix bundle consisting of α-helical folds
- Upon binding to its receptor, IL-10 activates signaling through JAK1 and STAT3
- It is produced by dendritic cells, macrophages, and CD4+ T cells, as well as mast cells, NK cells, neutrophils, and B cells, under specific stimulating conditions (Saraiva & O'Garra)
- IL-10 can inhibit the expression of pro-inflammatory cytokines and promote healing processes, and is important for the function of regulatory T cells
- IL-10 also enhances B cell proliferation, immunoglobulin secretion, and class II MHC expression, while IL-10 produced by macrophages inhibits activation of neighboring macrophages, thus allowing a level of self-regulation (Ouyang et al.)
Specifications
Dendritic Cells; Hematopoietic Stem and Progenitor Cells; Myeloid Cells; Neurons; T Cells | |
≥95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Human Recombinant IL-10