missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant TGF-beta 3
Transforming growth factor beta 3

2596.05 SEK - 68359.95 SEK
Specifications
Name | ARVD, FLJ16571, TGFB3, TGF-b3, Transforming growth factor beta-3 |
---|---|
Cell Type | Mesenchymal Cells, PSC-Derived; Mesenchymal Stem and Progenitor Cells |
Purity or Quality Grade | ≥95% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17109641
![]() |
STEMCELL Technologies
78131.2 |
4 x 250 ÎĽg |
68359.95 SEK
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17189631
![]() |
STEMCELL Technologies
78131 |
10 ÎĽg |
2596.05 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199631
![]() |
STEMCELL Technologies
78131.1 |
100 ÎĽg |
13701.15 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Transforming growth factor (TGF) beta 3 is a member of the TGF-β superfamily and regulates diverse cellular phenotypes
- TGF-beta 3 binds to serine-threonine kinase type I and II receptors and activates signal transduction through SMAD2/3 proteins, thus regulating a variety of functions, including cell proliferation, differentiation, wound healing, apoptosis, and metabolism (Massagué; McDowell et al.) TGF-beta 3 enhances glycosaminoglycan production by mesenchymal stromal cells, stimulates scar-free healing, and improves glucose tolerance and phenotypic changes in adipocyte morphology (Hall et al.; Holton et al.) TGF-beta 3 induces proliferation of posterofrontal suture-derived mesenchymal stromal cells, and stimulates expression of fibroblast growth factors 2 and 18 (James et al.)
Specifications
ARVD, FLJ16571, TGFB3, TGF-b3, Transforming growth factor beta-3 | |
Mesenchymal Cells, PSC-Derived; Mesenchymal Stem and Progenitor Cells | |
≥95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Human Recombinant TGF-beta 3