Learn More
Gibco™ Human IL-15 Receptor alpha (soluble) Fc Recombinant Protein, PeproTech®
Recombinant Protein
884.00 SEK - 12200.00 SEK
Specifications
Accession Number | Q13261 |
---|---|
For Use With (Application) | Functional Assay |
Format | Lyophilized |
Formulation | protein with no preservative |
Gene ID (Entrez) | 3601 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17819595
|
Gibco™
200-15RA-20UG |
20 μg |
884.00 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17829575
|
Gibco™
200-15RA-1MG |
1 mg |
12200.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17819665
|
Gibco™
200-15RA-500UG |
500 μg |
7270.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17809255
|
Gibco™
200-15RA-250UG |
250 μg |
4255.00 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17899535
|
Gibco™
200-15RA-100UG |
100 μg |
2175.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Recombinant Human sIL-15 Receptor alpha Fc has a calculated molecular weight of 44.4 kDa protein and contains 408 amino acid residues, which consists of the extracellular domain of IL-15R alpha fused to the Fc portion of human IgG. Due to glycosylation, it migrates at an apparent molecular weight of 50-60 kDa, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Interleukin 15 (IL-15RA) is a cytokine receptor that specifically binds IL-15 with high affinity. IL-15 regulates T and natural killer cell activation and proliferation. The IL-15 and IL-2 receptors share two subunits, IL-2R beta and IL-2R gamma, and IL-15RA is structurally related to IL-2RA, an additional IL-2-specific alpha subunit necessary for high affinity IL-2 binding. Unlike IL-2RA, IL-15RA is capable of binding IL-15 with high affinity independent of the other subunits. IL-15RA is thought to enhance cell proliferation and expression of apoptosis inhibitor Bcl-xL and Bcl-2.Specifications
Q13261 | |
Lyophilized | |
3601 | |
Human IL-15 Receptor alpha (soluble) Fc | |
RUO | |
AA690181; CD215; IL 15 receptor; IL 15 receptor subunit a; IL 15 R; IL 15 R alpha; IL 15 R a; IL15 receptor; IL15 receptor subunit alpha; IL-15 receptor subunit alpha; IL15 receptor subunit a; IL15R; IL15R alpha; IL15R a; Il15ra; IL-15 RA; IL-15 R-alpha; Interleukin 15 receptor; interleukin 15 receptor alpha isoform EM2; interleukin 15 receptor alpha isoform IC2; interleukin 15 receptor alpha isoform IC3; interleukin 15 receptor alpha isoform IC4; interleukin 15 receptor alpha isoform IC5; interleukin 15 receptor alpha isoform IC6; interleukin 15 receptor alpha isoform IC7; interleukin 15 receptor alpha isoform IC8; interleukin 15 receptor subunit alpha; Interleukin 15 receptor subunit a; interleukin 15 receptor, alpha; interleukin 15 receptor, alpha chain; interleukin-15 receptor subunit alpha; sIL 15 R; sIL-15 receptor subunit alpha; sIL15R; sIL-15 RA; sIL-15 R-alpha; soluble IL 15 R; soluble IL15R; Soluble interleukin-15 receptor subunit alpha | |
IL15RA | |
Protein | |
Recombinant | |
ITCPPPMSVE HADIWVKSYS LYSRERYICN SGFKRKAGTS SLTECVLNKA TNVAHWTTPS LKCIRDPALV HQRPAPPSTV TTAGVTPQPE SLSPSGKEPA ASSPSSNNTA ATTAAIVPGS QLMPSKSPST GTTEISSHES SHGTPSQTTA KNWELTASAS HQPPGVYPQG HSDTTGGPKS CDKTHTCPPC PAPELLGGPS VFLFPPKPKD TLMISRTPEV TCVVVDVSHE DPEVKFNWYV DGVEVHNAKT KPREEQYNST YRVVSVLTVL HQDWLNGKEY KCKVSNKALP APIEKTISKA KGQPREPQVY TLPPSRDELT KNQVSLTCLV KGFYPSDIAV EWESNGQPEN NYKTTPPVLD SDGSFFLYSK LTVDKSRWQQ GNVFSCSVMH EALHNHYTQK SLSLSPGK | |
Determined by its ability to inhibit the proliferation of mouse CTLL-2 cells in the presence of Recombinant Human IL-15. | |
Lyophilized |
Functional Assay | |
protein with no preservative | |
44.4 kDa | |
HPLC, SDS-PAGE | |
<1 EU/μg | |
IL15RA | |
Determined by its ability to inhibit the proliferation of mouse CTLL-2 cells in the presence of Recombinant Human IL-15. | |
Unconjugated | |
Fc-tag | |
-20°C | |
Insect cells | |
≥98% by SDS-PAGE gel and HPLC analyses. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.