Learn More
HYDRA-POINT™ Solvent G for two-component system, J.T. Baker™ Product Code.: 10211311
HYDRA-POINT™ Solvent G for two-component system, J.T. Baker™
Product Code. p-7220843 Shop All J.T.Baker Products
1 L, 1 L
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.50 L
Product Code. 10211311
Brand: JT Baker 8855.1000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Pyridine free solvent for volumetric Karl Fischer titration
  • Volumetric reagents for water determination by Karl Fischer titration
  • No matter what your requirements are for volumetric systems, HYDRA-POINT™ Karl Fischer reagents make water determination easy and trouble-free
  • They are pyridine-free and specially formulated for existing application procedures to give you consistent, reliable results for all types of samples
  • Formulated to reach each endpoint quickly, accurately and with reproducibility
  • Accuracy- reagents have titres that are more stable with less drift, helping you save time and reduce reagent usage
  • To be used with HYDRA-POINT™ Titrant 2 product no. 8845 or HYDRA-POINT™ Titrant 5 product no. 8844
Specifications
Quantity 1 L
Use Test Suitability for Karl Fischer titration: Passes Test
UN Number 1992
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydra-Point Solvent G
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H361-Suspected of damaging fertility or the unborn child.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : methanol, imidazole,sulphur dioxide
Product Content Correction
Product Title
HYDRA-POINT™ Solvent G for two-component system, J.T. Baker™ >