Learn More
Ansell HyFlex™ 11-132 Anti-Static Lint-Free Polyamide Gloves
Gloves are knitted using a seamless non-stop process with a textile liner made of copper-doped polyamide thread. Anti-static properties protect users from electrostatic discharge.
323.00 SEK
Specifications
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
---|---|
Construction | Seamless Knit |
Color | Natural |
Hand | Ambidextrous |
Material | Intercept Technology Yarn |
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | ||||||
16009591
|
Ansell
11132060 |
6 | N/A | |||||||
16019591
|
Ansell
11132070 |
7 | N/A | |||||||
16029591
|
Ansell
11132080 |
8 | N/A | |||||||
16039591
|
Ansell
11132090 |
9 | N/A | |||||||
16049591
|
Ansell
11132100 |
10 |
323.00 SEK
12 pairs Qty Price Break Available |
In Stock
Log in to see stock. |
||||||
Description
- Knitted with a seamless, non-stop process
- Elastic polyamide textile liner doped with copper
- Antistatic properties protect the user against electrostatic discharge
- Ambidextrous shape doubles the life of the glove by enabling the use of both sides
- Lint free gloves feature fine, stretchable stitches
Specifications
EN 420:2003 + A1:2009 | |
Natural | |
Intercept Technology Yarn | |
Knit |
Seamless Knit | |
Ambidextrous | |
1131A | |
11-132 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Ansell HyFlex™ 11-132 Anti-Static Lint-Free Polyamide Gloves