Learn More
IBA1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26177-1-AP-20UL
Description
The IBA1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human IBA1. This antibody recognizes human, mouse, rat antigen. The IBA1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
IBA1 | |
Polyclonal | |
Unconjugated | |
20 μL | |
199 | |
IgG |
Immunohistochemistry, ELISA | |
420 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.