Learn More
IBA1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26177-1-AP-20UL
 View more versions of this product
Product Code. 16884695
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The IBA1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human IBA1. This antibody recognizes human, mouse, rat antigen. The IBA1 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
Specifications
IBA1
Polyclonal
Unconjugated
20 μL
199
IgG
Immunohistochemistry, ELISA
420 μg/mL
Rabbit
Primary
Human, Rat, Mouse
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IBA1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 20 μL