missing translation for 'onlineSavingsMsg'
Learn More

IDO1 Mouse anti-Human, Mouse, Rat, Clone: 3G2G11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66528-1-IG-20UL

 View more versions of this product

Product Code. 16888485

  • 1645.54 SEK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

IDO1 is an intracellular heme-containing enzyme that catalyzes the oxidative cleavage of the indole ring of several important regulatory molecules like tryptophan, serotonin, and melatonin. By doing this, IDO1 initiates the production of biologically active metabolites, commonly referred to as kynurenines. IDO1 is widely expressed in a variety of human tissues as well as in macrophages and dendritic cells (DCs). In inflammation, interferons (IFNs) act on specific receptors to trigger IDO1 induction. The production of IFN-gamma and induction of IDO1 represent important antimicrobial mechanisms. Degradation and depletion of tryptophan by IDO1 inhibits the growth of viruses, bacteria and parasites. Furthermore, IDO1 plays a complex and crucial role in immunoregulation during infection, pregcy, autoimmunity, transplantation, and neoplasia.
Specifications

Specifications

IDO1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14902, P28776, Q9ERD9
Ido1
IDO1 Fusion Protein Ag3953
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG1
Western Blot
3G2G11
Unconjugated
IDO1
CD107B, IDO, IDO 1, IDO1, INDO, indoleamine 2,3 dioxygenase 1
Mouse
Protein G
RUO
15930, 3620, 66029
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IDO1 Mouse anti-Human, Mouse, Rat, Clone: 3G2G11, Proteintech > 20 μL; Unconjugated

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.