missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IER2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23849-1-AP-20UL
This item is not returnable.
View return policy
Description
DNA-binding protein that seems to act as a transcription factor. Involved in the regulation of neuronal differentiation, acts upon JNK-signaling pathway activation and plays a role in neurite outgrowth in hippocampal cells. May mediate with FIBP FGF-signaling in the establishment of laterality in the embryo. Promotes cell motility, seems to stimulate tumor metastasis.Specifications
IER2 | |
Polyclonal | |
Unconjugated | |
IER2 | |
3CH92, CHX1, ETR101, IER2, immediate early response 2, Pip92, Protein ETR101 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9592 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BTL4 | |
IER2 | |
IER2 Fusion Protein Ag20869 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IER2 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction