missing translation for 'onlineSavingsMsg'
Learn More

IER2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen IER2
Concentration 0.28 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16882475
View Documents
Proteintech
23849-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16872475
View Documents
Proteintech
23849-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

DNA-binding protein that seems to act as a transcription factor. Involved in the regulation of neuronal differentiation, acts upon JNK-signaling pathway activation and plays a role in neurite outgrowth in hippocampal cells. May mediate with FIBP FGF-signaling in the establishment of laterality in the embryo. Promotes cell motility, seems to stimulate tumor metastasis.
Specifications

Specifications

IER2
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
Q9BTL4
9592
IER2 Fusion Protein Ag20869
Primary
-20°C
IER2
0.28 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
3CH92, CHX1, ETR101, IER2, immediate early response 2, Pip92, Protein ETR101
IER2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IER2 Rabbit anti-Human, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.