Learn More
IFITM1-Specific Mouse anti-Human, Unconjugated, Clone: 5B5E2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 60074-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16897035
  • 1656.73 SEK / 20µL
Estimated Shipment: 22-05-2024
to see stock.
Description
Description
The IFITM1-Specific antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IFITM1-Specific. This antibody recognizes human antigen. The IFITM1-Specific antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
Specifications
IFITM1-Specific
Monoclonal
1500 μg/mL
Mouse
Primary
Human
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry
5B5E2
Unconjugated
20 μL
8519
IgG2a