Learn More
IFITM1-Specific Mouse anti-Human, Unconjugated, Clone: 5B5E2, Proteintech
Mouse Monoclonal Antibody
1662.46 SEK - 4700.76 SEK
Specifications
Antigen | IFITM1-Specific |
---|---|
Clone | 5B5E2 |
Concentration | 1500 ÎĽg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16897035
|
Proteintech
60074-1-IG-20UL |
20 ÎĽL |
1662.46 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16887035
|
Proteintech
60074-1-IG-150UL |
150 ÎĽL |
4700.76 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IFITM1-Specific antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IFITM1-Specific. This antibody recognizes human antigen. The IFITM1-Specific antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
IFITM1-Specific | |
1500 ÎĽg/mL | |
Monoclonal | |
Mouse | |
8519 | |
Primary |
5B5E2 | |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
Unconjugated | |
Human | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.