Learn More
IFITM1-Specific Mouse anti-Human, Unconjugated, Clone: 5B5E2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60074-1-IG-20UL
Description
The IFITM1-Specific antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IFITM1-Specific. This antibody recognizes human antigen. The IFITM1-Specific antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
IFITM1-Specific | |
Monoclonal | |
1500 ÎĽg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, Flow Cytometry | |
5B5E2 | |
Unconjugated | |
20 ÎĽL | |
8519 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.