Learn More
IFNAR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13083-1-AP-20UL
 View more versions of this product
Product Code. 16881124
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system. These cytokines produce their effects by binding to the Type 1 Interferon and Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1.
Specifications
Specifications
IFNAR1
Polyclonal
Unconjugated
IFNAR1
AVP, CRF2 1, IFN alpha REC, IFN alpha/beta receptor 1, IFN R 1, IFNAR, IFNAR1, IFNBR, IFRC, Type I interferon receptor 1
Rabbit
Antigen Affinity Chromatography
RUO
3454
-20°C
Liquid
Western Blot, Flow Cytometry
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P17181
IFNAR1
IFNAR1 Fusion Protein Ag3739
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IFNAR1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated