missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IFT81 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11744-1-AP-150UL
This item is not returnable.
View return policy
Description
Isoform CDV-1 may be involved in cardiac hypertrophy caused by carnitine deficiency. Isoform CDV-1R appears to play a role in development of the testis and spermatogenesis.Specifications
IFT81 | |
Polyclonal | |
Unconjugated | |
IFT81 | |
CDV 1, CDV 1R, CDV1, CDV1R, IFT81 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100856715, 12589, 28981 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O35594, Q8WYA0 | |
IFT81 | |
IFT81 Fusion Protein Ag2339 | |
150 μL | |
Primary | |
Canine, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IFT81 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction