missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL11RA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10264-1-AP-150UL
This item is not returnable.
View return policy
Description
Interleukin 11 is a stromal cell-derived cytokine that belongs to a family of pleiotropic and redundant cytokines that use the gp130 transducing subunit in their high affinity receptors. This gene encodes the IL-11 receptor, which is a member of the hematopoietic cytokine receptor family. This particular receptor is very similar to ciliary neurotrophic factor, since both contain an extracellular region with a 2-domain structure composed of an immunoglobulin-like domain and a cytokine receptor-like domain. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
IL11RA | |
Polyclonal | |
Unconjugated | |
IL11RA | |
IL 11 receptor subunit alpha, IL 11R alpha, IL 11R subunit alpha, IL 11RA, IL11RA, interleukin 11 receptor, alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16157, 245983, 3590 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q14626, Q64385, Q99MF4 | |
IL11RA, Il11ra1 | |
IL11RA Fusion Protein Ag0373 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IL11RA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction