missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL11RA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | IL11RA |
---|---|
Concentration | 0.55 mg/mL |
Applications | Immunofluorescence, Flow Cytometry, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16896533
|
Proteintech
10264-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16886533
|
Proteintech
10264-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Interleukin 11 is a stromal cell-derived cytokine that belongs to a family of pleiotropic and redundant cytokines that use the gp130 transducing subunit in their high affinity receptors. This gene encodes the IL-11 receptor, which is a member of the hematopoietic cytokine receptor family. This particular receptor is very similar to ciliary neurotrophic factor, since both contain an extracellular region with a 2-domain structure composed of an immunoglobulin-like domain and a cytokine receptor-like domain. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
IL11RA | |
Immunofluorescence, Flow Cytometry, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q14626, Q64385, Q99MF4 | |
16157, 245983, 3590 | |
IL11RA Fusion Protein Ag0373 | |
Primary | |
-20°C | |
IL11RA |
0.55 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
IL 11 receptor subunit alpha, IL 11R alpha, IL 11R subunit alpha, IL 11RA, IL11RA, interleukin 11 receptor, alpha | |
IL11RA, Il11ra1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IL11RA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech