missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL28B/ IFNL3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24199-1-AP-150UL
This item is not returnable.
View return policy
Description
The interleukins are a broad family of well characterized cytokines, primarily of hematopoietic cell origin. As new cytokines are molecularly characterized, they are assigned an IL number to maintain a standard nomenclature. The interleukins are secreted by immune cells (mainly macrophages, B-cells or T-cells) that regulate a wide range of immune system functions. The functions of different interleukins vary from the regulation of inflammatory and immune responses to the regulation of other interleukins. IL-28 and IL-29 are induced by viral infection and show antiviral activity. IL-28 may play a role in antiviral immunity through up-regulation of MHC class I antigen expression. IL-28 is a ligand for the heterodimeric class II cytokine receptor composed of IL10RB and IL28RA.Specifications
IL28B/ IFNL3 | |
Polyclonal | |
Unconjugated | |
IFNL3 | |
Cytokine Zcyto22, IFN lambda 3, IFN lambda 4, IFNL3, IFNL4, IL 28B, IL 28C, IL28B, IL28C, Interferon lambda 3, Interferon lambda 4, Interleukin 28B, Interleukin 28C, ZCYTO22 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
282617 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IZI9 | |
IFNL3 | |
IL28B/ IFNL3 Fusion Protein Ag21491 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IL28B/ IFNL3 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction