Learn More
Imide-extended bismaleimide, powder, BMI-3000, Thermo Scientific™
Product Code. 15432747
100 g
Click to view available options
Quantity:
25 g
100 g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 921213-77-6
Specifications
Color Yellow
Physical Form Powder
Quantity 100 g
Chemical Name or Material Imide-extended bismaleimide, BMI-3000
Product Content Correction
Product Title
Imide-extended bismaleimide, powder, BMI-3000, Thermo Scientific™ >