Learn More
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection
Electrode cables to connect Mettler Toledo InLab sensors to any Mettler Toledo meter.
Brand: Mettler Toledo™ 30281928
1759.02 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Detachable
- Resuable
Specifications
S7-None | |
To connect Mettler Toledo InLab sensors to any meter | |
10 m |
Mettler Toledo InLab Sensors | |
32.8 ft. |
Product Content Correction
Product Title
Mettler Toledo™ InLab™ Sensor Cables: S7 Connection > Connector: None; Length: 10m (32.8 ft.)
Spot an opportunity for improvement?Share a Content Correction