missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INPP5D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | INPP5D |
---|---|
Concentration | 0.26 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899164
|
Proteintech
19694-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889164
|
Proteintech
19694-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to INPP5D.
an SH2-containing inositol phosphatase. A hemopoietic-specific phosphatase that regulates cell survival, growth, cell cycle arrest and apoptosis. Hydrolyzes Ins(1, 3, 4, 5)P4 and PtdIns(3, 4, 5)P3. A cytosolic protein with a SH2 domain in its N-terminus and two NPXY Shc binding motifs at its C-terminus. Upon receptor cross-linking, SHIP is first recruited to the plasma membrane by its SH2, followed by tyrosine phosphorylation on the NPXY motif. Both membrane relocalization and phosphorylation are essential for its regulatory function. Associates with SHC after cytokine stimulation.Specifications
INPP5D | |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P97573, Q92835, Q9ES52 | |
16331, 3635, 54259 | |
Peptide | |
Primary | |
-20°C | |
INPP5D |
0.26 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hp51CN, INPP5D, p150Ship, SHIP, SHIP 1, SHIP1, SIP 145 | |
INPP5D | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
INPP5D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech