Learn More
INPP5D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19694-1-AP-150UL
 View more versions of this product
Product Code. 16889164
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This antibody is specific to INPP5D.
an SH2-containing inositol phosphatase. A hemopoietic-specific phosphatase that regulates cell survival, growth, cell cycle arrest and apoptosis. Hydrolyzes Ins(1, 3, 4, 5)P4 and PtdIns(3, 4, 5)P3. A cytosolic protein with a SH2 domain in its N-terminus and two NPXY Shc binding motifs at its C-terminus. Upon receptor cross-linking, SHIP is first recruited to the plasma membrane by its SH2, followed by tyrosine phosphorylation on the NPXY motif. Both membrane relocalization and phosphorylation are essential for its regulatory function. Associates with SHC after cytokine stimulation.
Specifications
Specifications
INPP5D
Polyclonal
Unconjugated
INPP5D
hp51CN, INPP5D, p150Ship, SHIP, SHIP 1, SHIP1, SIP 145
Rabbit
Antigen Affinity Chromatography
RUO
16331, 3635, 54259
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P97573, Q92835, Q9ES52
INPP5D
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
INPP5D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated