Learn More
Iron Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
684.00 SEK - 1410.00 SEK
Specifications
Solution Contains | HNO3 |
---|---|
Concentration | 1 g/L |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12986184
![]() |
Fisher Chemical
S 119 100 |
100 mL |
684.00 SEK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12996184
![]() |
Fisher Chemical
S 119 250 |
250 mL |
1100.00 SEK
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12906194
![]() |
Fisher Chemical
S 119 500 |
500 mL |
1410.00 SEK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
HNO3 | |
1 g/L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ICP Standard Solution Iron (Fe) 1000mg/l in HNO
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands, forearms andface thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : iron trinitrate, nitric acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Iron Solution for ICP, 1g/L, HNO3 Matrix, Fisher Chemical™