missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIN17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12313-1-AP-20UL
This item is not returnable.
View return policy
Description
KIN17 has a calculated molecular weight of 45 kDa, this antibody is raised against human KIN17 and detects a band of about 40 kDa.
Kin is a nuclear protein that forms intranuclear foci during proliferation and is redistributed in the nucleoplasm during the cell cycle. Short-wave ultraviolet light provokes the relocalization of the protein, suggesting its participation in the cellular response to DNA damage. Originally selected based on protein-binding with RecA antibodies, the mouse protein presents a limited similarity with a functional domain of the bacterial RecA protein, a characteristic shared by the human ortholog.Specifications
KIN17 | |
Polyclonal | |
Unconjugated | |
KIN | |
Binding to curved DNA, BTCD, DNA/RNA binding protein KIN17, KIN, KIN17 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16588, 22944, 689197 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60870, Q8K339 | |
Kin | |
KIN17 Fusion Protein Ag2963 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KIN17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction