missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIN17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | KIN17 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899953
|
Proteintech
12313-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889953
|
Proteintech
12313-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KIN17 has a calculated molecular weight of 45 kDa, this antibody is raised against human KIN17 and detects a band of about 40 kDa.
Kin is a nuclear protein that forms intranuclear foci during proliferation and is redistributed in the nucleoplasm during the cell cycle. Short-wave ultraviolet light provokes the relocalization of the protein, suggesting its participation in the cellular response to DNA damage. Originally selected based on protein-binding with RecA antibodies, the mouse protein presents a limited similarity with a functional domain of the bacterial RecA protein, a characteristic shared by the human ortholog.Specifications
KIN17 | |
Immunocytochemistry, Immunoprecipitation, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
O60870, Q8K339 | |
16588, 22944, 689197 | |
KIN17 Fusion Protein Ag2963 | |
Primary | |
-20°C | |
KIN |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Binding to curved DNA, BTCD, DNA/RNA binding protein KIN17, KIN, KIN17 | |
Kin | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KIN17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech