missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHDC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27277-1-AP-20UL
This item is not returnable.
View return policy
Description
KLHDC1 contains 6 Kelch repeats and is widely expressed, with high levels in skeletal muscle, pancreas and liver. KLHDC1 and KLHDC2 have differential localization and activity in cultured mammalian cells. The exact function of KLHDC1 remains unknown.Specifications
KLHDC1 | |
Polyclonal | |
Unconjugated | |
KLHDC1 | |
kelch domain containing 1, KLHDC1, MST025 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
122773, 271005, 314190 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q80YG3, Q8N7A1 | |
Klhdc1 | |
KLHDC1 Fusion Protein Ag23704 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
KLHDC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction