missing translation for 'onlineSavingsMsg'
Learn More

KLRB1/CD161 Mouse anti-Human, Unconjugated, Clone: 3D8F5, Proteintech

Mouse Monoclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen KLRB1/CD161
Clone 3D8F5
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16840226
View Documents Promotion Details
Proteintech
67537-1-IG-20UL
20 μL
1656.73 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16830226
View Documents Promotion Details
Proteintech
67537-1-IG-150UL
150 μL
4721.67 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 18-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The KLRB1/CD161 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KLRB1/CD161. This antibody recognizes human antigen. The KLRB1/CD161 antibody has been validated for the following applications: IHC, WB, ELISA analysis.

Specifications

Specifications

KLRB1/CD161
1000 μg/mL
Monoclonal
Mouse
3820
Primary
3D8F5
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Human
IgG1
Videos
SDS
Documents

Documents

Product Certifications