missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Dehydroascorbic acid, 96%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 228.0°C |
Assay Percent Range | 95.0 % min. (ELSD) (HPLC) |
Quantity | 1 g |
Formula Weight | 174.11 |
Chemical Name or Material | L-Dehydroascorbic acid |
Safety and Handling

Product Identifier
- L-Dehydroascorbic acid
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : L-Dehydroascorbic acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Dehydroascorbic acid, 96% >
Spot an opportunity for improvement?Share a Content Correction