missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tryptophan, ≥99%, MP Biomedicals™
Shop All MP Biomedical Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
5g
Specifications
Specifications
Color | White |
Assay Percent Range | ≥99% |
Quantity | 5 g |
Formula Weight | 204.22 |
Physical Form | Crystalline Powder |
Chemical Name or Material | L-Tryptophan |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Tryptophan, ≥99%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction