missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Glove Box Moisture Monitor

Moisture Monitor, 0 - 6000 ppm, for Labconco Glove Boxes

77990.00 SEK

Specifications

For Use With (Application) Measures dew point range from -80 to 0°C or 0 to 6000ppm
View More Specs

Products 1
Product Code Brand Voltage Price Quantity & Availability  
Product Code Brand Voltage Price Quantity & Availability  
17164504
View Documents
Labconco™
5442201
230 V
77990.00 SEK
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Moisture monitor measures dew point range from -80 to 0° C or 0 to 6000 ppm. Displays dew point only. Includes meter, probe, attachment plug, and calibration stand. For placement inside the glove box’s main chamber. For use with for Labconco Protector and Precise Controlled Atmosphere and Combination Glove Boxes.
Specifications

Specifications

Measures dew point range from -80 to 0°C or 0 to 6000ppm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco™ Glove Box Moisture Monitor

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.