Learn More
LAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 16136-1-AP-150UL
 View more versions of this product
Product Code. 16885654
  • 4652.90 SEK / 150µL
Description
Immunohistochemical staining of 16136-1-AP shows strong extracellular matrixpositivity in human testis. The predicted MW of this protein is 57 kDa. IP results of 16136-1-AP detected a 62 kDa band.
Anchoring filament protein which is a component of the basement membrane zone.
Specifications
Specifications
LAD1
Polyclonal
Unconjugated
LAD1
LAD, Lad 1, LAD1, LadA, ladinin 1, Linear IgA disease antigen
Rabbit
Antigen Affinity Chromatography
RUO
16763, 3898
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O00515, P57016
LAD1
LAD1 Fusion Protein Ag9159
150 μL
Primary
Human, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
LAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated