missing translation for 'onlineSavingsMsg'
Learn More

LAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen LAD1
Concentration 0.31 mg/mL
Applications Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16895654
View Documents
Proteintech
16136-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16885654
View Documents
Proteintech
16136-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Immunohistochemical staining of 16136-1-AP shows strong extracellular matrixpositivity in human testis. The predicted MW of this protein is 57 kDa. IP results of 16136-1-AP detected a 62 kDa band.

Anchoring filament protein which is a component of the basement membrane zone.
Specifications

Specifications

LAD1
Immunoprecipitation, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Mouse, Human
O00515, P57016
16763, 3898
LAD1 Fusion Protein Ag9159
Primary
-20°C
LAD1
0.31 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
LAD, Lad 1, LAD1, LadA, ladinin 1, Linear IgA disease antigen
LAD1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
LAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.