missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDLRAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20096-1-AP-20UL
This item is not returnable.
View return policy
Description
The antibody can recognize all 3 isoforms of LDLRAD1.
LDLRAD1 belongs to the LDLR family.Specifications
LDLRAD1 | |
Polyclonal | |
Unconjugated | |
LDLRAD1 | |
LDLRAD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
388633, 546840 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5T700 | |
LDLRAD1 | |
Peptide | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LDLRAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction