missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDLRAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | LDLRAD1 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879574
|
Proteintech
20096-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869574
|
Proteintech
20096-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody can recognize all 3 isoforms of LDLRAD1.
LDLRAD1 belongs to the LDLR family.Specifications
LDLRAD1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q5T700 | |
388633, 546840 | |
Peptide | |
Primary | |
-20°C | |
LDLRAD1 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
LDLRAD1 | |
LDLRAD1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LDLRAD1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech