Läs mer
Brand™ LDPE Wash Bottles without Venting Valve
High-quality wash bottles with integrated wash stream and sealing cone closure.
Brand: Brand™ 144080
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
- For distilled water and organic solvents only
- Large opening for convenient filling
- Label with designation of contents and general safety statements in English (was CAS numbers and NFPA codes) and German (was CAS numbers)
- Manufactured from flexible PE-LD, limited to temperatures below 80°C
Specifikationer
| 250 mL | |
| Green | |
| Wash Bottle |
| LDPE | |
| Wide | |
| Methanol |
Produktförslag
Korrigering av produktinnehåll
Produkttitel
Brand™ LDPE Wash Bottles without Venting Valve > Chemical label: Methanol, Capacity: 250mL
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering