Learn More
Brand™ LDPE Wash Bottles without Venting Valve
High-quality wash bottles with integrated wash stream and sealing cone closure.
317.00 SEK
Specifications
Capacity (Metric) | 250 mL |
---|---|
Material | LDPE |
Mouth | Wide |
Product Type | Wash Bottle |
Description
- For distilled water and organic solvents only
- Large opening for convenient filling
- Label with designation of contents and general safety statements in English (was CAS numbers and NFPA codes) and German (was CAS numbers)
- Manufactured from flexible PE-LD, limited to temperatures below 80°C
Specifications
250 mL | |
LDPE | |
Wide | |
Wash Bottle |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Brand™ LDPE Wash Bottles without Venting Valve