missing translation for 'onlineSavingsMsg'
Learn More
Novus Biologicals™ Lightning-Link (R) APC Antibody Labeling Kit Product Code.: 18725553

Novus Biologicals™ Lightning-Link (R) APC Antibody Labeling Kit

Product Code. 18725553 Shop All Bio Techne Products
1 mg, 1mg
Click to view available options
Quantity:
1 mg
100 ÎĽg
30 ÎĽg
300 ÎĽg
Unit Size:
1 set
100µg
1mg
300µg
This item is not returnable. View return policy

Product Code. 18725553

Brand: Novus Biologicals™ 7050015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody labeling kit

Allophycocyanin is a large 105kDa fluorescent phycobiliprotein commonly used in FACS analysis. It absorbs at 650nm and emits at 662nm.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 1 mg
Conjugate APC (Allophycocyanin)
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Product Type APC Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Lightning-Link (R) APC Antibody Labeling Kit >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.