Learn More
Novus Biologicals™ Lightning-Link (R) APC Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Unit Size:
1 set
100µg
1mg
300µg
Description
Allophycocyanin is a large 105kDa fluorescent phycobiliprotein commonly used in FACS analysis. It absorbs at 650nm and emits at 662nm.
Order Info
Quantity
Specifications
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
Quantity | 30 μg |
Conjugate | APC (Allophycocyanin) |
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent |
For Use With (Application) | Flow Cytometry, Immunocytochemistry/Immunofluorescence |
Product Type | APC Antibody Labeling Kit |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Novus Biologicals™ Lightning-Link (R) APC Antibody Labeling Kit >
Spot an opportunity for improvement?Share a Content Correction