missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
1 x 100 μg Reaction
3 x 0.01 mg Reaction
300 μg
Unit Size:
1 set
100µg
1mg
30µg
Description
APC/Cy7 is a tandem conjugate. In the Lightning-Link APC/Cy7 the tandem is excited at 633nm, wavelength at which the APC has its maximum absorption. The excited APC functions then as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy7. The Cy7 emits then the energy received from the APC in the form of long wavelength light at 776nm.
Order Info
Quantity

Specifications
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
Quantity | 300 μg |
Conjugate | APC/Cy7 |
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent |
Product Type | APC-Cy7 Antibody Labeling Kit |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit >
Spot an opportunity for improvement?Share a Content Correction