missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) PerCP-Cy5.5 Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Unit Size:
1 set
100µg
1mg
Description
PerCP/Cy5.5 is a tandem conjugate. The PerCP is excited at 482nm and functions as an energy donor for the Cy5.5. Energy is transferred from the PerCP to the Cy5.5 via energy resonance transfer. The Cy5.5 emits the energy received from the PerCP in the form of long wavelength light at 700nm.
Order Info
Quantity

Specifications
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
Quantity | 100 μg |
Conjugate | PerCP/Cy5.5 |
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent |
Product Type | PerCP-Cy5.5 Antibody Labeling Kit |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Lightning-Link (R) PerCP-Cy5.5 Antibody Labeling Kit >
Spot an opportunity for improvement?Share a Content Correction