missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) PerCP-Cy5.5 Antibody Labeling Kit

Applications: Flow Cytometry, Electron Microscopy, Flow (Cell Surface)

Brand:  Novus Biologicals™ 763-0015

 View more versions of this product

Product Code. 18774973

  • 11035.00 SEK / 1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PerCP/Cy5.5 is a tandem conjugate. The PerCP is excited at 482nm and functions as an energy donor for the Cy5.5. Energy is transferred from the PerCP to the Cy5.5 via energy resonance transfer. The Cy5.5 emits the energy received from the PerCP in the form of long wavelength light at 700nm.

Order Info

Quantity

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at -20°C. Avoid freeze-thaw cycles.
PerCP/Cy5.5
PerCP-Cy5.5 Antibody Labeling Kit
1 mg
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Lightning-Link (R) PerCP-Cy5.5 Antibody Labeling Kit > 1mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.