missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) PerCP-Cy5.5 Antibody Labeling Kit
Applications: Flow Cytometry, Electron Microscopy, Flow (Cell Surface)
4340.00 SEK - 11035.00 SEK
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
---|---|
Conjugate | PerCP/Cy5.5 |
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent |
Product Type | PerCP-Cy5.5 Antibody Labeling Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18725883
|
Novus Biologicals™
763-0005 |
100 ÎĽg |
4340.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18757553
|
Novus Biologicals™
763-0010 |
300 ÎĽg |
7730.00 SEK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18779663
|
Novus Biologicals™
763-0030 |
30 ÎĽg |
5080.00 SEK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18774973
|
Novus Biologicals™
763-0015 |
1 mg |
11035.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PerCP/Cy5.5 is a tandem conjugate. The PerCP is excited at 482nm and functions as an energy donor for the Cy5.5. Energy is transferred from the PerCP to the Cy5.5 via energy resonance transfer. The Cy5.5 emits the energy received from the PerCP in the form of long wavelength light at 700nm.Specifications
Store at -20°C. Avoid freeze-thaw cycles. | |
PerCP/Cy5.5 | |
Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent | |
PerCP-Cy5.5 Antibody Labeling Kit |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Lightning-Link (R) PerCP-Cy5.5 Antibody Labeling Kit