missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN7A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25150-1-AP-150UL
This item is not returnable.
View return policy
Description
Plays a role in establishing and maintaining the asymmetric distribution of channels and receptors at the plasma membrane of polarized cells. Forms membrane-associated multiprotein complexes that may regulate delivery and recycling of proteins to the correct membrane domains. The tripartite complex composed of LIN7 (LIN7A, LIN7B or LIN7C), CASK and APBA1 may have the potential to couple synaptic vesicle exocytosis to cell adhesion in brain. Ensures the proper localization of GRIN2B (subunit 2B of the NMDA receptor) to neuronal postsynaptic density and may function in localizing synaptic vesicles at synapses where it is recruited by beta-catenin and cadherin. Required to localize Kir2 channels, GABA transporter (SLC6A12) and EGFR/ERBB1, ERBB2, ERBB3 and ERBB4 to the basolateral membrane of epithelial cells.Specifications
LIN7A | |
Polyclonal | |
Unconjugated | |
LIN7A | |
hLin 7, lin 7 homolog A (C. elegans), LIN 7A, LIN7, LIN7A, MALS 1, MALS1, Mammalian lin seven protein 1, Protein lin 7 homolog A, Tax interaction protein 33, TIP 33, Veli 1, VELI1, Vertebrate lin 7 homolog 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
108030, 85327, 8825 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14910, Q8JZS0, Q9Z250 | |
Lin7a | |
LIN7A Fusion Protein Ag18440 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LIN7A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction