missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN7A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | LIN7A |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883695
|
Proteintech
25150-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873695
|
Proteintech
25150-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plays a role in establishing and maintaining the asymmetric distribution of channels and receptors at the plasma membrane of polarized cells. Forms membrane-associated multiprotein complexes that may regulate delivery and recycling of proteins to the correct membrane domains. The tripartite complex composed of LIN7 (LIN7A, LIN7B or LIN7C), CASK and APBA1 may have the potential to couple synaptic vesicle exocytosis to cell adhesion in brain. Ensures the proper localization of GRIN2B (subunit 2B of the NMDA receptor) to neuronal postsynaptic density and may function in localizing synaptic vesicles at synapses where it is recruited by beta-catenin and cadherin. Required to localize Kir2 channels, GABA transporter (SLC6A12) and EGFR/ERBB1, ERBB2, ERBB3 and ERBB4 to the basolateral membrane of epithelial cells.Specifications
LIN7A | |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O14910, Q8JZS0, Q9Z250 | |
108030, 85327, 8825 | |
LIN7A Fusion Protein Ag18440 | |
Primary | |
-20°C | |
LIN7A |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
hLin 7, lin 7 homolog A (C. elegans), LIN 7A, LIN7, LIN7A, MALS 1, MALS1, Mammalian lin seven protein 1, Protein lin 7 homolog A, Tax interaction protein 33, TIP 33, Veli 1, VELI1, Vertebrate lin 7 homolog 1 | |
Lin7a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LIN7A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech