missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20678-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
VAMP7 is a member of the soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) family, localizing to late endosomes and lysosomes. VAMP7 is thought to mediate the fusion of endosomes to their target lysosomes as well as other exocytosis events during phagocytosis and neuritogenesis. VAMP7 interacts with the VPS9 ankyrin repeat protein VARP, a protein that localizes to early endosomes and thought to regulate endosome dynamics. Together with CD82, VAMP7 can modulate the signaling of EGFR by regulating its endocytosis from the plasma membrane.Specifications
LIS1 | |
Polyclonal | |
Unconjugated | |
PAFAH1B1 | |
LIS 1, LIS1, LIS2, Lissencephaly 1 protein, MDCR, MDS, PAF AH 45 kDa subunit, PAF AH alpha, PAFAH, PAFAH alpha, PAFAH1B1, PAFAHA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18472, 5048, 83572 | |
-20°C | |
Liquid |
Western Blot | |
0.24 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P43034, P63004, P63005 | |
Pafah1b1 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |