Learn More
Eppendorf™ Locking Clip
Prevent spring action of tip cones with these locking clips
Brand: Eppendorf™ 3122612006
80.00 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory for Eppendorf Xplorer™/Xplorer plus
- Locking clip
- Available for 8- and 12-channel pipettes
Specifications
Locking Clip | |
Preventing spring action of tip cones | |
8-channel pipette | |
1 Each |
Promotions
-
Multiple Manufacturers
Genomics Powerhouse
Power your pipeline from PCR to sequencing.Promo Code: 24117Expires: 2025-08-24
New Offer Save Up to 92%
Product Content Correction
Product Title
Eppendorf™ Locking Clip > For 8-channel Pipet
Spot an opportunity for improvement?Share a Content Correction